6-Chloroisatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione
OR400007-1G
OR400007-5G
OR400007-25G
APOSOR400007-1GEA
49.3
EUR
APOSOR400007-1G
APOSOR400007-5G
APOSOR400007-25G
6-Chloroisatoic anhydride
6-Chloroisatoic anhydride
Formel:
C₈H₄ClNO₃ Molecular Weight: 197,575 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09032504 CAS-Nummer: 20829-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...