5-Methoxyisatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Methoxy-1H-benzo[d][1,3]oxazine-2,4-dione, 6-Methoxy-2H-3,1-benzoxazine-2,4(1H)-dione
OR400011-250MG
APOSOR400011-250MGEA
21.9
EUR
APOSOR400011-250MG
5-Methoxyisatoic anhydride
5-Methoxyisatoic anhydride
Formel:
C₉H₇NO₄ Molecular Weight: 193,156 g/mol Schmelzpunkt: 244…246 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179411 CAS-Nummer: 37795-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...