2,2-Dimethyl-4-oxobutanenitrile 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2-Dimethyl-4-oxobutyronitrile, 3-Cyano-3-methylbutanal

OR40004-100MG OR40004-250MG
APOSOR40004-100MGEA 157 EUR
APOSOR40004-100MG APOSOR40004-250MG
2,2-Dimethyl-4-oxobutanenitrile 95%
2,2-Dimethyl-4-oxobutanenitrile
Formel: C₆H₉NO
Molecular Weight: 111,142 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07437928
CAS-Nummer: 18240-73-8
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR