2-Bromo-5-iodophenylacetic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR400042-1G
OR400042-250MG
APOSOR400042-1GEA
806
EUR
APOSOR400042-1G
APOSOR400042-250MG
2-Bromo-5-iodophenylacetic acid
2-Bromo-5-iodophenylacetic acid
Formel:
C₈H₆BrIO₂ Molecular Weight: 340,941 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18391762 CAS-Nummer: 1261680-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...