4-Nitro-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

OR400059-1G OR400059-5G
APOSOR400059-1GEA 40.6 EUR
APOSOR400059-1G APOSOR400059-5G
4-Nitro-1,3-benzoxazole
4-Nitro-1,3-benzoxazole
Formel: C₇H₄N₂O₃
Molecular Weight: 164,118 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00965006
CAS-Nummer: 163808-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR