5-Iodo-2-nitroanisole
Lieferant: Apollo Scientific
|
Achtung
|
OR400064-1G
OR400064-5G
APOSOR400064-1GEA
92.8
EUR
APOSOR400064-1G
APOSOR400064-5G
5-Iodo-2-nitroanisole
5-Iodo-2-nitroanisole
Formel:
C₇H₆INO₃ Schmelzpunkt: 92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18397934 CAS-Nummer: 860585-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...