4-Bromo-3-iodobenzoic acid

Lieferant: Apollo Scientific
Achtung

OR400071-1G OR400071-25G OR400071-5G
APOSOR400071-1GEA 21.9 EUR
APOSOR400071-1G APOSOR400071-25G APOSOR400071-5G
4-Bromo-3-iodobenzoic acid
4-Bromo-3-iodobenzoic acid
Formel: C₇H₄BrIO₂
Molecular Weight: 326,914 g/mol
Schmelzpunkt: 243…245 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12406813
CAS-Nummer: 42860-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR