2-Bromo-5-hydroxybenzoic acid

Lieferant: Apollo Scientific
Achtung

OR400079-1G OR400079-25G OR400079-5G
APOSOR400079-1GEA 36.4 EUR
APOSOR400079-1G APOSOR400079-25G APOSOR400079-5G
2-Bromo-5-hydroxybenzoic acid
2-Bromo-5-hydroxybenzoic acid
Formel: C₇H₅BrO₃
Molecular Weight: 217,017 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797974
CAS-Nummer: 58380-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR