Methyl 2-bromo-5-hydroxybenzoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-bromo-5-hydroxybenzenecarboxylate
OR400090-1G
OR400090-5G
OR400090-25G
APOSOR400090-1GEA
109
EUR
APOSOR400090-1G
APOSOR400090-5G
APOSOR400090-25G
Methyl 2-bromo-5-hydroxybenzoate
Methyl 2-bromo-5-hydroxybenzoate
Formel:
C₈H₇BrO₃ Molecular Weight: 231,043 g/mol Schmelzpunkt: 96…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10566797 CAS-Nummer: 154607-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...