Methyl 5-bromo-2-methoxybenzoate
Lieferant: Apollo Scientific
|
Achtung
|
OR400107-1G
OR400107-25G
APOSOR400107-1GEA
0
EUR
APOSOR400107-1G
APOSOR400107-25G
Methyl 5-bromo-2-methoxybenzoate
Methyl 5-bromo-2-methoxybenzoate
Formel:
C₉H₉BrO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203656 CAS-Nummer: 7120-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...