2-Bromo-6-iodobenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

OR400119-250MG OR400119-1G
APOSOR400119-250MGEA 219 EUR
APOSOR400119-250MG APOSOR400119-1G
2-Bromo-6-iodobenzyl alcohol 95%
2-Bromo-6-iodobenzyl alcohol
Formel: C₇H₆BrIO
Molecular Weight: 312.93 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11520412
CAS-Nummer: 1261850-41-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR