2-Bromo-6-iodobenzyl bromide 95+%

Lieferant: Apollo Scientific
Gefahr

OR400121-250MG OR400121-1G OR400121-5G
APOSOR400121-250MGEA 21.9 EUR
APOSOR400121-250MG APOSOR400121-1G APOSOR400121-5G
2-Bromo-6-iodobenzyl bromide 95+%
2-Bromo-6-iodobenzyl bromide
Formel: C₇H₅Br₂I
Molecular Weight: 375,827 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391668
CAS-Nummer: 1261498-12-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR