2'-Bromo-6'-iodoacetophenone 95%

Lieferant: Apollo Scientific
Achtung

OR400125-250MG OR400125-1G
APOSOR400125-250MGEA 172 EUR
APOSOR400125-250MG APOSOR400125-1G
2'-Bromo-6'-iodoacetophenone 95%
2'-Bromo-6'-iodoacetophenone
Formel: C₈H₆BrIO
Molecular Weight: 324,941 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391608
CAS-Nummer: 1261438-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR