4-Iodo-3-nitrobenzonitrile 95%

Lieferant: Apollo Scientific

OR400133-10G OR400133-1G OR400133-5G
APOSOR400133-10GEA 202 EUR
APOSOR400133-10G APOSOR400133-1G APOSOR400133-5G
4-Iodo-3-nitrobenzonitrile 95%
4-Iodo-3-nitrobenzonitrile
Formel: C₇H₃IN₂O₂
Molecular Weight: 274.015 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07783655
CAS-Nummer: 101420-79-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR