2-Chloro-6-methoxyaniline

Lieferant: Apollo Scientific

OR400164-1G OR400164-5G
APOSOR400164-1GEA 55.1 EUR
APOSOR400164-1G APOSOR400164-5G
2-Chloro-6-methoxyaniline
2-Chloro-6-methoxyaniline
Formel: C₇H₈ClNO
Molecular Weight: 157.598 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14455911
CAS-Nummer: 158966-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR