2-Chloro-5-iodobenzaldehyde

Lieferant: Apollo Scientific
Achtung

OR400186-1G OR400186-250MG OR400186-5G
APOSOR400186-1GEA 324 EUR
APOSOR400186-1G APOSOR400186-250MG APOSOR400186-5G
2-Chloro-5-iodobenzaldehyde
2-Chloro-5-iodobenzaldehyde
Formel: C₇H₄ClIO
Molecular Weight: 266,464 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12024988
CAS-Nummer: 256925-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR