4-Iodo-5-methyl-2-nitroaniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-iodo-4-methylnitrobenzene, 5-Amino-2-iodo-4-nitrotoluene, 4-Iodo-6-nitro-m-toluidine
OR400191-1G
OR400191-250MG
OR400191-5G
APOSOR400191-1GEA
685
EUR
APOSOR400191-1G
APOSOR400191-250MG
APOSOR400191-5G
4-Iodo-5-methyl-2-nitroaniline
4-Iodo-5-methyl-2-nitroaniline
Formel:
C₇H₇IN₂O₂ Molecular Weight: 278.047 g/mol Schmelzpunkt: 166…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD27949584 CAS-Nummer: 335349-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...