3-Bromo-5-methylbenzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Bromo-5-methylphenyl)methanol, 3-Bromo-5-methylbenzyl Alcohol
OR400195-1G
OR400195-250MG
OR400195-5G
APOSOR400195-1GEA
63.8
EUR
APOSOR400195-1G
APOSOR400195-250MG
APOSOR400195-5G
3-Bromo-5-methylbenzyl alcohol
3-Bromo-5-methylbenzyl alcohol
Formel:
C₈H₉BrO Molecular Weight: 201,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18204791 CAS-Nummer: 648439-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...