4-Bromo-5-methyl-2-nitroanisole 95+%
Lieferant: Apollo Scientific
Synonyme:
1-Bromo-4-methoxy-2-methyl-5-nitrobenzene
OR400208-1G
OR400208-5G
APOSOR400208-1GEA
60.9
EUR
APOSOR400208-1G
APOSOR400208-5G
4-Bromo-5-methyl-2-nitroanisole 95+%
4-Bromo-5-methyl-2-nitroanisole
Formel:
C₈H₈BrNO₃ Molecular Weight: 246,058 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23703052 CAS-Nummer: 89978-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...