3-(Methylsulphonyl)benzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Formylphenyl methyl sulphone
OR40021-1G
OR40021-5G
OR40021-250MG
APOSOR40021-1GEA
43.5
EUR
APOSOR40021-1G
APOSOR40021-5G
APOSOR40021-250MG
3-(Methylsulphonyl)benzaldehyde
3-(Methylsulphonyl)benzaldehyde
Formel:
C₈H₈O₃S Molecular Weight: 184,212 g/mol Schmelzpunkt: 88,7…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11052329 CAS-Nummer: 43114-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...