2-Amino-5-iodophenol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-4-iodoaniline

OR400213-1G OR400213-250MG OR400213-5G
APOSOR400213-1GEA 161 EUR
APOSOR400213-1G APOSOR400213-250MG APOSOR400213-5G
2-Amino-5-iodophenol 95%
2-Amino-5-iodophenol
Formel: C₆H₆INO
Molecular Weight: 235,022 g/mol
Schmelzpunkt: 126…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD14705905
CAS-Nummer: 99968-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR