3-[(Trimethylsilyl)ethynyl]benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

OR400242-250MG OR400242-1G OR400242-5G
APOSOR400242-250MGEA 29 EUR
APOSOR400242-250MG APOSOR400242-1G APOSOR400242-5G
3-[(Trimethylsilyl)ethynyl]benzoic acid 97%
3-[(Trimethylsilyl)ethynyl]benzoic acid
Formel: C₁₂H₁₄O₂Si
Molecular Weight: 218,324 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28040529
CAS-Nummer: 16116-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR