4-Bromo-2-chloro-6-iodophenol 95%

Lieferant: Apollo Scientific
Gefahr

OR400330-1G OR400330-250MG OR400330-5G
APOSOR400330-1GEA 348 EUR
APOSOR400330-1G APOSOR400330-250MG APOSOR400330-5G
4-Bromo-2-chloro-6-iodophenol 95%
4-Bromo-2-chloro-6-iodophenol
Formel: C₆H₃BrClIO
Molecular Weight: 333,349 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26895843
CAS-Nummer: 858855-18-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR