(4S)-2,2-Dioxido-4- benzyl -1,2,3-oxathiazolidine, N-BOC protected 95%

Lieferant: Apollo Scientific
Achtung

OR400337-1G OR400337-250MG OR400337-5G
APOSOR400337-1GEA 232 EUR
APOSOR400337-1G APOSOR400337-250MG APOSOR400337-5G
(4S)-2,2-Dioxido-4- benzyl -1,2,3-oxathiazolidine, N-BOC protected 95%
(4S)-2,2-Dioxido-4- benzyl -1,2,3-oxathiazolidine, N-BOC protected
Formel: C₁₄H₁₉NO₅S
Lagertemperatur: Kühlschrank
MDL: MFCD28122848
CAS-Nummer: 1105712-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR