2-Vinylthiophene 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR400376-1G
OR400376-250MG
OR400376-5G
APOSOR400376-1GEA
50.9
EUR
APOSOR400376-1G
APOSOR400376-250MG
APOSOR400376-5G
2-Vinylthiophene 97%
2-Vinylthiophene
Formel:
C₆H₆S Lagertemperatur: Kühlschrank |
MDL:
MFCD00085296 CAS-Nummer: 1918-82-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...