5-Chloro-3-iodoisatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-8-iodo-1H-3,1-benzoxazine-2,4-dione
OR400378-1G
OR400378-250MG
OR400378-5G
APOSOR400378-1GEA
232
EUR
APOSOR400378-1G
APOSOR400378-250MG
APOSOR400378-5G
5-Chloro-3-iodoisatoic anhydride
5-Chloro-3-iodoisatoic anhydride
Formel:
C₈H₃ClINO₃ Molecular Weight: 323,472 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28154805 CAS-Nummer: 33115-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...