2-Bromo-4-methoxy-6-nitroaniline
Lieferant: Apollo Scientific
|
Achtung
|
OR400396-1G
OR400396-25G
OR400396-5G
APOSOR400396-1GEA
26.1
EUR
APOSOR400396-1G
APOSOR400396-25G
APOSOR400396-5G
2-Bromo-4-methoxy-6-nitroaniline
2-Bromo-4-methoxy-6-nitroaniline
Formel:
C₇H₇BrN₂O₃ Molecular Weight: 247.046 g/mol Schmelzpunkt: 119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18088521 CAS-Nummer: 10172-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...