4-Amino-3-iodophenol

Lieferant: Apollo Scientific
Achtung

OR400417-1G OR400417-250MG
APOSOR400417-1GEA 364 EUR
APOSOR400417-1G APOSOR400417-250MG
4-Amino-3-iodophenol
4-Amino-3-iodophenol
Formel: C₆H₆INO
Molecular Weight: 235.022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16999396
CAS-Nummer: 66416-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR