2-Bromo-3-chloroanisole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-chloro-3-methoxybenzene

OR400506-1G OR400506-250MG OR400506-5G
APOSOR400506-1GEA 29 EUR
APOSOR400506-1G APOSOR400506-250MG APOSOR400506-5G
2-Bromo-3-chloroanisole
2-Bromo-3-chloroanisole
Formel: C₇H₆BrClO
Molecular Weight: 221,479 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18390300
CAS-Nummer: 174913-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR