Methyl 3-amino-4-bromobenzo[b]thiophene-2-carboxylate
Lieferant: Apollo Scientific
OR400509-1G
OR400509-250MG
APOSOR400509-1GEA
328
EUR
APOSOR400509-1G
APOSOR400509-250MG
Methyl 3-amino-4-bromobenzo[b]thiophene-2-carboxylate
Methyl 3-amino-4-bromobenzo[b]thiophene-2-carboxylate
Formel:
C₁₀H₈BrNO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD28348430 CAS-Nummer: 1980045-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...