2-Bromo-3-nitrobenzyl alcohol 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR400519-1G
OR400519-250MG
OR400519-5G
APOSOR400519-1GEA
281
EUR
APOSOR400519-1G
APOSOR400519-250MG
APOSOR400519-5G
2-Bromo-3-nitrobenzyl alcohol 95%
2-Bromo-3-nitrobenzyl alcohol
Formel:
C₇H₆BrNO₃ Molecular Weight: 232,031 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18392283 CAS-Nummer: 90407-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...