4-Iodo-3-methoxybenzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR400604-1G
OR400604-250MG
APOSOR400604-1GEA
993
EUR
APOSOR400604-1G
APOSOR400604-250MG
4-Iodo-3-methoxybenzyl bromide 95%
4-Iodo-3-methoxybenzyl bromide
Formel:
C₈H₈BrIO Molecular Weight: 326,957 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18905029 CAS-Nummer: 1379341-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...