5-Cyano-2-hydroxybenzoic acid

Lieferant: Apollo Scientific
Achtung

OR400616-100MG OR400616-1G OR400616-250MG
APOSOR400616-100MGEA 155 EUR
APOSOR400616-100MG APOSOR400616-1G APOSOR400616-250MG
5-Cyano-2-hydroxybenzoic acid
5-Cyano-2-hydroxybenzoic acid
Formel: C₈H₅NO₃
Molecular Weight: 163,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16999178
CAS-Nummer: 10435-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR