2-Bromo-3-nitroaniline

Lieferant: Apollo Scientific
Gefahr

OR400621-1G OR400621-5G OR400621-250MG
APOSOR400621-1GEA 133 EUR
APOSOR400621-1G APOSOR400621-5G APOSOR400621-250MG
2-Bromo-3-nitroaniline
2-Bromo-3-nitroaniline
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12923157
CAS-Nummer: 35757-20-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR