4-Bromo-2-iodo-6-nitrophenol

Lieferant: Apollo Scientific
Achtung

OR400632-1G OR400632-250MG OR400632-5G
APOSOR400632-1GEA 313 EUR
APOSOR400632-1G APOSOR400632-250MG APOSOR400632-5G
4-Bromo-2-iodo-6-nitrophenol
4-Bromo-2-iodo-6-nitrophenol
Formel: C₆H₃BrINO₃
Molecular Weight: 343,901 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28397822
CAS-Nummer: 58349-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR