5-Bromo-2-nitrobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromo-2-nitrophenyl)methanol

OR400635-1G OR400635-5G OR400635-25G
APOSOR400635-1GEA 21.9 EUR
APOSOR400635-1G APOSOR400635-5G APOSOR400635-25G
5-Bromo-2-nitrobenzyl alcohol
5-Bromo-2-nitrobenzyl alcohol
Formel: C₇H₆BrNO₃
Molecular Weight: 232,031 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848688
CAS-Nummer: 1241894-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR