4-Iodo-3-methoxybenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

OR400639-1G OR400639-250MG
APOSOR400639-1GEA 406 EUR
APOSOR400639-1G APOSOR400639-250MG
4-Iodo-3-methoxybenzyl alcohol 95%
4-Iodo-3-methoxybenzyl alcohol
Formel: C₈H₉IO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09909621
CAS-Nummer: 244257-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR