2-Bromo-5-methoxybenzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-5-methoxybenzene-1-sulphonyl chloride

OR400647-1G OR400647-250MG
APOSOR400647-1GEA 87 EUR
APOSOR400647-1G APOSOR400647-250MG
2-Bromo-5-methoxybenzenesulphonyl chloride
2-Bromo-5-methoxybenzenesulphonyl chloride
Formel: C₇H₆BrClO₃S
Molecular Weight: 285,543 g/mol
Schmelzpunkt: 62…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09880303
CAS-Nummer: 179251-57-1
UN: 2928
ADR: 6.1+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR