4’-Bromo-3’-methylacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-3-methylphenyl)ethanone

OR400663-1G OR400663-5G
APOSOR400663-1GEA 26.1 EUR
APOSOR400663-1G APOSOR400663-5G
4’-Bromo-3’-methylacetophenone
4’-Bromo-3’-methylacetophenone
Formel: C₉H₉BrO
Molecular Weight: 213,071 g/mol
Schmelzpunkt: 31 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045017
CAS-Nummer: 37074-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR