3-Chloro-2-methoxybenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR400666-1G
OR400666-250MG
APOSOR400666-1GEA
473
EUR
APOSOR400666-1G
APOSOR400666-250MG
3-Chloro-2-methoxybenzonitrile
3-Chloro-2-methoxybenzonitrile
Formel:
C₈H₆ClNO Molecular Weight: 167,592 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393570 CAS-Nummer: 725718-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...