5-Bromo-3-iodo-2-methoxyaniline

Lieferant: Apollo Scientific
Achtung

OR400723-1G OR400723-250MG
APOSOR400723-1GEA 905 EUR
APOSOR400723-1G APOSOR400723-250MG
5-Bromo-3-iodo-2-methoxyaniline
5-Bromo-3-iodo-2-methoxyaniline
Formel: C₇H₇BrINO
Molecular Weight: 327,945 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28898256
CAS-Nummer: 1845713-72-5
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR