2-Bromo-4-hydroxybenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR400738-1G OR400738-250MG OR400738-5G
APOSOR400738-1GEA 49.3 EUR
APOSOR400738-1G APOSOR400738-250MG APOSOR400738-5G
2-Bromo-4-hydroxybenzoic acid 95%
2-Bromo-4-hydroxybenzoic acid
Formel: C₇H₅BrO₃
Molecular Weight: 217,017 g/mol
Schmelzpunkt: 206…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16999959
CAS-Nummer: 28547-28-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR