3-Bromo-2-iodonitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-iodo-3-nitrobenzene
OR400777-1G
OR400777-250MG
OR400777-5G
APOSOR400777-1GEA
68.3
EUR
APOSOR400777-1G
APOSOR400777-250MG
APOSOR400777-5G
3-Bromo-2-iodonitrobenzene
3-Bromo-2-iodonitrobenzene
Formel:
C₆H₃BrINO₂ Molecular Weight: 327,902 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16660176 CAS-Nummer: 32337-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...