3-Bromo-5-formylbenzoic acid 96%

Lieferant: Apollo Scientific
Achtung

OR400779-1G OR400779-5G OR400779-10G
APOSOR400779-1GEA 78.3 EUR
APOSOR400779-1G APOSOR400779-5G APOSOR400779-10G
3-Bromo-5-formylbenzoic acid 96%
3-Bromo-5-formylbenzoic acid
Formel: C₈H₅BrO₃
Molecular Weight: 229.027 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22544371
CAS-Nummer: 398119-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR