2-Bromo-4-iodobenzonitrile

Lieferant: Apollo Scientific
Achtung

OR400783-1G OR400783-250MG OR400783-5G
APOSOR400783-1GEA 24.8 EUR
APOSOR400783-1G APOSOR400783-250MG APOSOR400783-5G
2-Bromo-4-iodobenzonitrile
2-Bromo-4-iodobenzonitrile
Formel: C₇H₃BrIN
Molecular Weight: 307,914 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391643
CAS-Nummer: 1261776-03-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR