2-Bromo-3-chloro-6-hydroxybenzonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
OR400797-250MG
OR400797-1G
OR400797-5G
APOSOR400797-250MGEA
23.2
EUR
APOSOR400797-250MG
APOSOR400797-1G
APOSOR400797-5G
2-Bromo-3-chloro-6-hydroxybenzonitrile
2-Bromo-3-chloro-6-hydroxybenzonitrile
Formel:
C₇H₃BrClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD29037276 CAS-Nummer: 1934463-16-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...