3-Amino-5-chloropyrazine-2-carbonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR40081-1G
OR40081-250MG
APOSOR40081-1GEA
420
EUR
APOSOR40081-1G
APOSOR40081-250MG
3-Amino-5-chloropyrazine-2-carbonitrile 95%
3-Amino-5-chloropyrazine-2-carbonitrile
Formel:
C₅H₃ClN₄ Molecular Weight: 154,557 g/mol Schmelzpunkt: 194…196 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD10697779 CAS-Nummer: 54632-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...