1-Hydroxy-4-nitroisoquinoline 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Nitroisoquinolin-1(2H)-one

OR400882-1G OR400882-250MG OR400882-5G
APOSOR400882-1GEA 60.9 EUR
APOSOR400882-1G APOSOR400882-250MG APOSOR400882-5G
1-Hydroxy-4-nitroisoquinoline 97%
1-Hydroxy-4-nitroisoquinoline
Formel: C₉H₆N₂O₃
Molecular Weight: 190.156 g/mol
Schmelzpunkt: 247…248 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03861155
CAS-Nummer: 55404-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR