6-Bromo-3-iodo-2-nitroanisole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-iodo-2-methoxy-3-nitrobenzene

OR400889-1G OR400889-250MG OR400889-5G
APOSOR400889-1GEA 1220 EUR
APOSOR400889-1G APOSOR400889-250MG APOSOR400889-5G
6-Bromo-3-iodo-2-nitroanisole
6-Bromo-3-iodo-2-nitroanisole
Formel: C₇H₅BrINO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD30730277
CAS-Nummer: 2149590-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR