4-Bromo-1,3-benzoxazole 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromobenzo[d]oxazole
OR40093-1G
OR40093-250MG
APOSOR40093-1GEA
166
EUR
APOSOR40093-1G
APOSOR40093-250MG
4-Bromo-1,3-benzoxazole 98+%
4-Bromo-1,3-benzoxazole
Formel:
C₇H₄BrNO Molecular Weight: 198,017 g/mol Schmelzpunkt: 53…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234620 CAS-Nummer: 217326-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...